Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pentadecanoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Pentadecylic acidn-Pentadecanoic acid

A14664-18 A14664-30 A14664-18 A14664-30 A14664-09 A14664-09
AAAA14664-09EA 70.77 USD InStock
CAAAAA14664-30 AAAA14664-09 CAAAAA14664-09 AAAA14664-18 CAAAAA14664-18 AAAA14664-30
Pentadecanoic acid 99%
Pentadecanoic acid


Formula: C₁₅H₃₀O₂
MW: 242.4 g/mol
Boiling Pt: 257 °C/100 mm
Melting Pt: 51…54 °C
Density: 0,8496 g/cm³ (70 °C)
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002745
CAS Number: 1002-84-2
EINECS: 213-693-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty