Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylbiphenyl 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methyl-1,1'-biphenyl4-Phenyltoluene

A14702-03 A14702-03 A14702-14 A14702-14 A14702-06 A14702-06
AAAA14702-06EA 121.99 USD InStock
AAAA14702-06 AAAA14702-03 AAAA14702-14 CAAAAA14702-06 CAAAAA14702-03 CAAAAA14702-14
4-Methylbiphenyl 98%
4-Methylbiphenyl


Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 44…48 °C
Density: 1.015
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008544
CAS Number: 644-08-6
EINECS: 211-409-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty