Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Ethoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14758-14 A14758-36 A14758-14 A14758-22 A14758-22 A14758-36
AAAA14758-14EA 29.99 USD InStock
AAAA14758-14 AAAA14758-36 CAAAAA14758-36 CAAAAA14758-14 AAAA14758-22 CAAAAA14758-22
2-Ethoxybenzoic acid 98%
2-Ethoxybenzoic acid


Formula: C₉H₁₀O₃
Boiling Pt: 174…175 °C/15 mm
Melting Pt: 19…20 °C
Density: 1,105
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002438
CAS Number: 134-11-2
EINECS: 205-130-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty