Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dibromobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A14765-22 A14765-22 A14765-18 A14765-30 A14765-14 A14765-30 A14765-14 A14765-18
AAAA14765-14EA 85.09 USD InStock
AAAA14765-14 CAAAA14765-18 AAA14765-18 CAAAAA14765-22 AAAA14765-22 CAAAAA14765-14 CAAAAA14765-30 AAAA14765-30
1,2-Dibromobenzene 98%
1,2-Dibromobenzene


Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 224…226 °C
Melting Pt: 4…6 °C
Density: 1.985
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000057
CAS Number: 583-53-9
EINECS: 209-507-3
UN: 3082
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty