Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4,5-dichloroimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-4,5-dichloro-1H-imidazole

A14896-03 A14896-06 A14896-06 A14896-14 A14896-14 A14896-03
AAAA14896-14EA 864.71 USD InStock
CAAAAA14896-03 CAAAAA14896-14 CAAAAA14896-06 AAAA14896-14 AAAA14896-03 AAAA14896-06
2-Bromo-4,5-dichloroimidazole 98%
2-Bromo-4,5-dichloroimidazole


Formula: C₃HBrCl₂N₂
MW: 215.86 g/mol
Melting Pt: ca 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00067894
CAS Number: 16076-27-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty