Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Heptafluorobutyric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Heptafluorobutanoic acid Perfluorobutyric acid HFBA

A14907-22 A14907-22 A14907-14 A14907-36 A14907-36 A14907-14
AAAA14907-36EA 770.5 USD InStock
AAAA14907-36 CAAAAA14907-36 AAA14907-14 AAA14907-22 CAAAA14907-14 CAAAA14907-22
Heptafluorobutyric acid 99%
Heptafluorobutyric acid


Formula: C₄HF₇O₂
MW: 214.04 g/mol
Boiling Pt: 120…121 °C
Melting Pt: –18 °C
Density: 1.648
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00004171
CAS Number: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty