Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triphosgene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bis(trichloromethyl) carbonate

A14932-22 A14932-14 A14932-06
AAAA14932-14EA 179.56 USD InStock
AAAA14932-14 AAAA14932-22 AAAA14932-06
Triphosgene 98%
Triphosgene


Formula: C₃Cl₆O₃
MW: 296.75 g/mol
Boiling Pt: 203…206 °C
Melting Pt: 77…91 °C
Density: 1.600
Storage Temperature: Refrigerator
MDL Number: MFCD00062848
CAS Number: 32315-10-9
EINECS: 250-986-3
UN: 2928
ADR: "6.1",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty