Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl levulinate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Oxopentanoic acid ethyl ester 4-Oxovaleric acid ethyl ester Levulinic acid ethyl ester 4-Ketovaleric acid ethyl ester Ethyl 4-oxovalerate

A15001-18 A15001-18 A15001-36 A15001-36 A15001-22 A15001-22
AAA15001-18EA 29.29 USD InStock
CAAAAA15001-22 AAA15001-18 AAAA15001-36 AAAA15001-22 CAAAAA15001-36 CAAAA15001-18
Ethyl levulinate 98%
Ethyl levulinate


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 203…205 °C
Density: 1.014
Flash Pt: 94 °C(201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009209
CAS Number: 539-88-8
EINECS: 208-728-2
Merck Index: 13,03852
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty