Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(4-Carboxyphenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(2-Carboxyethyl)benzoic Acid

A15081-06 A15081-09 A15081-14 A15081-06 A15081-09 A15081-22 A15081-22 A15081-14
AAA15081-06EA 107.99 USD InStock
AAA15081-06 AAAA15081-22 AAA15081-14 CAAAAA15081-09 CAAAA15081-06 CAAAA15081-14 AAAA15081-09 CAAAAA15081-22
3-(4-Carboxyphenyl)propionic acid 98%
3-(4-Carboxyphenyl)propionic acid


Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 288…291 °C
Storage Temperature: Ambient
MDL Number: MFCD00016560
CAS Number: 38628-51-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty