Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt dihydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

A15161-0I A15161-30 A15161-0B A15161-30 A15161-0I A15161-0B
AAAA15161-30EA 77.34 USD InStock
AAAA15161-30 CAAAAA15161-30 AAAA15161-0B CAAAAA15161-0B AAAA15161-0I CAAAAA15161-0I
EDTA disodium salt dihydrate 99+%
EDTA disodium salt dihydrate


Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/56/2299956.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty