Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-4-fluorobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

A15198-14 A15198-06 A15198-06 A15198-14
AAAA15198-14EA 141.99 USD InStock
CAAAAA15198-14 AAAA15198-14 CAAAAA15198-06 AAAA15198-06
3-Bromo-4-fluorobenzaldehyde 98%
3-Bromo-4-fluorobenzaldehyde


Formula: C₇H₄BrFO
MW: 203.01 g/mol
Boiling Pt: 138…139 °C/2.5 mm
Melting Pt: 29…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042186
CAS Number: 77771-02-9
EINECS: 278-764-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty