Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-2-methoxyethane 96% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromoethyl methyl ether

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

A15370-36 A15370-22 A15370-14 A15370-14 A15370-06 A15370-36 A15370-06 A15370-22
AAAA15370-06EA 27 USD InStock
AAAA15370-06 AAAA15370-14 AAAA15370-36 CAAAAA15370-06 CAAAAA15370-22 CAAAAA15370-36 CAAAAA15370-14 AAAA15370-22
1-Bromo-2-methoxyethane 96% stabilized
1-Bromo-2-methoxyethane


Formula: BrH₂CCH₂OCH₃
MW: 138.99 g/mol
Boiling Pt: 109…110 °C
Density: 1.478
Flash Pt: 28 °C(82 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000236
CAS Number: 6482-24-2
EINECS: 229-339-4
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty