Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (4-Fluorophenyl)methanamine

A15486-30 A15486-30 A15486-09 A15486-18 A15486-18 A15486-09
AAAA15486-18EA 113.99 USD InStock
CAAAAA15486-18 AAAA15486-18 CAAAAA15486-09 AAAA15486-09 CAAAAA15486-30 AAAA15486-30
4-Fluorobenzylamine 98+%
4-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 182…183 °C
Density: 1.095
Flash Pt: 66 °C(150 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008120
CAS Number: 140-75-0
EINECS: 205-430-4
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty