Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium hydroxide monohydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15519-0E A15519-0E A15519-36 A15519-22 A15519-22 A15519-36
AAAA15519-22EA 66.99 USD InStock
AAAA15519-22 CAAAAA15519-36 CAAAAA15519-0E AAAA15519-36 CAAAAA15519-22 AAAA15519-0E
Lithium hydroxide monohydrate 98%
Lithium hydroxide monohydrate


Formula: LiOH·1H₂O
MW: 41.96 g/mol
Melting Pt: 462 °C
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: "8",II
Merck Index: 13,05556
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty