Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Butynoic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A15542-06 A15542-14 A15542-14 A15542-06
AAAA15542-14EA 625.99 USD InStock
CAAAAA15542-14 CAAAAA15542-06 AAAA15542-14 AAAA15542-06
2-Butynoic acid 98%
2-Butynoic acid


Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 200…203 °C
Melting Pt: 75…79 °C
Density: 0.964
Storage Temperature: Refrigerator
MDL Number: MFCD00004363
CAS Number: 590-93-2
EINECS: 209-695-7
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty