Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

A15623-18 A15623-06 A15623-06 A15623-09 A15623-09 A15623-18
AAAA15623-06EA 71.09 USD InStock
AAAA15623-06 CAAAA15623-09 CAAAAA15623-06 CAAAA15623-18 AAA15623-18 AAA15623-09
2-Fluorobenzylamine 97%
2-Fluorobenzylamine


Formula: C₇H₈FN
MW: 125.15 g/mol
Boiling Pt: 80…82 °C/18 mm
Density: 1.099
Flash Pt: 67 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008107
CAS Number: 89-99-6
EINECS: 201-957-9
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty