Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzyl chloroformate 95% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: CBZ-Cl (Z-Cl)

Stabilization: Stabilised with sodium carbonate (Na₂CO₃) ∼0.1%

A15682-18 A15682-30 A15682-30 A15682-0B A15682-0B
AAAA15682-30EA 227.71 USD InStock
CAAAAA15682-30 CAAAA15682-0B AAAA15682-30 CAAAA15682-18 AAA15682-0B
Benzyl chloroformate 95% stabilized
Benzyl chloroformate


Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 178…180 °C
Melting Pt: –20 °C
Density: 1.195
Flash Pt: 125 °C(257 °F)
Storage Temperature: Freezer
MDL Number: MFCD00000640
CAS Number: 501-53-1
EINECS: 207-925-0
UN: 1739
ADR: "8",I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty