Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzenesulfonamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15689-22 A15689-06 A15689-22 A15689-14 A15689-14 A15689-09 A15689-06 A15689-09
AAAA15689-22EA 567.99 USD InStock
AAAA15689-22 AAA15689-06 CAAAAA15689-22 CAAAA15689-06 CAAAA15689-14 AAAA15689-09 CAAAAA15689-09 AAA15689-14
2-Nitrobenzenesulfonamide 98+%
2-Nitrobenzenesulphonamide


Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00009807
CAS Number: 5455-59-4
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty