Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Anisic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methoxybenzoic acid 4-Anisic acid

A15697-0B A15697-30 A15697-0I A15697-0B A15697-0I A15697-30
AAA15697-30EA 53.89 USD InStock
CAAAA15697-0B AAA15697-30 CAAAAA15697-0I AAA15697-0B CAAAA15697-30 AAAA15697-0I
p-Anisic acid 98+%
p-Anisic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 275…277 °C
Melting Pt: 182…185 °C
Density: 1.385
Flash Pt: 185 °C(365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty