Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Butyne-1-ol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Hydroxy-2-butyne But-2-yn-1-ol

A15709-06 A15709-14 A15709-06 A15709-14 A15709-22
AAA15709-14EA 178.99 USD InStock
CAAAA15709-06 AAA15709-14 AAA15709-06 CAAAA15709-14 AAAA15709-22
2-Butyne-1-ol 98%
2-Butyne-1-ol


Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 142…143 °C
Melting Pt: –2 °C
Density: 0.938
Flash Pt: 51 °C(123 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002914
CAS Number: 764-01-2
EINECS: 212-113-4
UN: 1987
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty