Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Crotonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: trans-Crotonic acid 2-Butenoic acid trans-2-Butenoic acid

A15765-14 A15765-36 A15765-14 A15765-0E A15765-0E A15765-36
AAA15765-0EEA 224.99 USD InStock
AAA15765-0E AAA15765-36 AAAA15765-14 CAAAA15765-36 CAAAAA15765-14 CAAAA15765-0E
Crotonic acid 98%
Crotonic acid


Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 128 °C/100 mm
Melting Pt: 70…74 °C
Density: 1.027
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002701
CAS Number: 107-93-7
EINECS: 203-533-9
UN: 2823
ADR: "8",III
Merck Index: 13,02625
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty