Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diphenyl ether 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Diphenyl oxidePhenyl etherPhenoxybenzene

A15791-30 A15791-36 A15791-36 A15791-30 A15791-0E A15791-0E
AAAA15791-30EA 48.1 USD InStock
AAAA15791-30 CAAAAA15791-30 CAAAAA15791-36 AAAA15791-0E CAAAAA15791-0E AAAA15791-36
Diphenyl ether 99%
Diphenyl ether


Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 258…260 °C
Melting Pt: 26…29 °C
Density: 1.071
Flash Pt: 125 °C(257 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003034
CAS Number: 101-84-8
EINECS: 202-981-2
UN: 3077
ADR: "9",III
Merck Index: 12,07442
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty