Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenylbenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Biphenylylaldehyde p-Biphenylylaldehyde 4-Biphenylcarbaldehyde 4-Biphenylcarboxaldehyde Biphenyl-4-carboxaldehyde Biphenyl-4-carbaldehyde

A15839-14 A15839-06 A15839-22 A15839-14 A15839-22 A15839-06
AAA15839-14EA 125.99 USD InStock
CAAAA15839-14 AAA15839-14 AAAA15839-06 AAA15839-22 CAAAAA15839-06 CAAAA15839-22
4-Phenylbenzaldehyde 98+%
4-Phenylbenzaldehyde


Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 184 °C/11 mm
Melting Pt: 57…61 °C
Density: 1,107
Flash Pt: 172 °C(341 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006947
CAS Number: 3218-36-8
EINECS: 221-742-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty