Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phthaloylglycine 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

A15920-18 A15920-0B A15920-0B A15920-30 A15920-30 A15920-18
AAA15920-18EA 58.59 USD InStock
AAA15920-18 CAAAA15920-30 CAAAA15920-18 CAAAAA15920-0B AAAA15920-0B AAA15920-30
N-Phthaloylglycine 98+%
N-Phthaloylglycine


Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 194…198 °C
Storage Temperature: Ambient
MDL Number: MFCD00005900
CAS Number: 4702-13-0
EINECS: 225-177-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty