Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-tert-Butyl-1-bromobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Bromo-4-(tert-butyl)benzene2-(4-Bromophenyl)-2-methylpropane4-Bromo-tert-butylbenzene1-Bromo-4-tert-butylbenzene

A15924-14 A15924-36 A15924-36 A15924-22 A15924-14 A15924-22
AAA15924-14EA 59.09 USD InStock
CAAAA15924-14 AAA15924-14 CAAAAA15924-36 CAAAA15924-22 AAAA15924-36 AAA15924-22
4-tert-Butyl-1-bromobenzene 97%
4-tert-Butyl-1-bromobenzene


Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Boiling Pt: 231…232 °C
Melting Pt: 15…16 °C
Density: 1,229
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000108
CAS Number: 3972-65-4
EINECS: 223-599-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty