Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Propionaldehyde 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Propanal

A16146-AU A16146-0J A16146-AU A16146-AP A16146-AP A16146-0J
AAA16146-AUEA 61.85 USD InStock
AAA16146-AU AAAA16146-AP CAAAA16146-0J CAAAA16146-AU CAAAAA16146-AP AAA16146-0J
Propionaldehyde 97%
Propionaldehyde


Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 47…49 °C
Melting Pt: –81 °C
Density: 0.81
Flash Pt: -40 °C(-40 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: "3",II
Merck Index: 13,07915
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty