Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzenesulfonic acid 94%

Supplier: Thermo Scientific Chemicals
Danger

A16148-36 A16148-36 A16148-0E A16148-22
AAAA16148-36EA 38.99 USD InStock
AAAA16148-36 AAAA16148-0E AAA16148-22 CAAAAA16148-36
Benzenesulfonic acid 94%
Benzenesulphonic acid
Acid catalyst for direct esterification of amino acids and peptides


Formula: C₆H₆O₃S
MW: 158.18 g/mol
Melting Pt: ca 45…50 °C
Density: 1.32
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: "8",III
Merck Index: 13,01070
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty