Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-bromobenzoate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Bromobenzoic acid methyl ester

A16174-18 A16174-18 A16174-09 A16174-30 A16174-09 A16174-30
AAA16174-09EA 66.89 USD InStock
AAA16174-09 AAA16174-18 CAAAA16174-09 CAAAAA16174-30 AAAA16174-30 CAAAA16174-18
Methyl-3-bromobenzoate 98+%
Methyl 3-bromobenzoate


Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Boiling Pt: 139…140 °C/35 mm
Melting Pt: 30…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017777
CAS Number: 618-89-3
EINECS: 210-569-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty