Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Nonane 99%

Supplier: Thermo Scientific Chemicals
Danger

A16177-AP A16177-AE A16177-AP
AAAA16177-APEA 357.96 USD InStock
CAAAAA16177-AP AAAA16177-AP AAAA16177-AE
n-Nonane 99%
n-Nonane


Formula: H₃C(CH₂)₇CH₃
MW: 128.26 g/mol
Boiling Pt: 150…152 °C
Melting Pt: –53 °C
Density: 0.718
Flash Pt: 31 °C(87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009574
CAS Number: 111-84-2
EINECS: 203-913-4
UN: 1920
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty