Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2R,3R)-2,3-Bis(benzoyloxy)succinic acid

A16181-14 A16181-22 A16181-36 A16181-36 A16181-22 A16181-14
AAA16181-22EA 89.59 USD InStock
AAA16181-22 AAA16181-36 CAAAA16181-22 CAAAA16181-36 AAAA16181-14 CAAAAA16181-14
(-)-Dibenzoyl-L-tartaric acid, anhydrous 99%
(-)-Dibenzoyl-L-tartaric acid


Formula: C₁₈H₁₄O₈
MW: 358.31 g/mol
Melting Pt: 154…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00003074
CAS Number: 2743-38-6
EINECS: 220-374-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty