Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorbic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (E,E)-2,4-Hexadienoic acid2,4-Hexadienoic acid(2E,4E)-2,4-Hexadienoic acidtrans,trans-2,4-Hexadienoic acid(2E,4E)-Hexa-2,4-dienoic acid

A16196-30 A16196-0B A16196-0I A16196-36
AAAA16196-0BEA 107.99 USD InStock
AAAA16196-0B AAAA16196-36 AAAA16196-30 AAAA16196-0I
Sorbic acid 99%
Sorbic acid


Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228 °C (dec.)
Melting Pt: 132…135 °C
Density: 1.205
Flash Pt: 127 °C(260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://us-legacy.vwr.com/stibo/web/std.lang.all/29/27/2302927.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty