Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) chloride hexahydrate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Iron trichloride hexahydrateIron (III) chloride 6-hydrateFerric chloride hexahydrateIron trichloride 6-hydrateFerric chloride 6-hydrate

A16231-0C A16231-36 A16231-0C A16231-0E A16231-36 A16231-0E
AAAA16231-36EA 82.38 USD InStock
AAAA16231-36 AAAA16231-0C CAAAAA16231-0E AAAA16231-0E CAAAAA16231-0C CAAAAA16231-36
Iron(III) chloride hexahydrate 97%
Iron (III) chloride hexahydrate


Formula: FeCl₃·6H₂O
MW: 270.3 g/mol
Boiling Pt: 280…285 °C
Melting Pt: 37 °C
Density: 1.820
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
EINECS: 231-729-4
UN: 3260
ADR: "8",III
Merck Index: 13,04048
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/51/16640551.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty