Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Methylamino)phenol sulfate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Methylamino)phenol hemisulfate saltMetol4-(Methylamino)phenol hemisulphatep-(Methylamino)phenol sulphate4-(Methylamino)phenol hemisulfate4-Methylaminophenolsulphatep-(Methylamino)phenol sulfate4-(Methylamino)phenol hemisulphate salt4-Methylaminophenolsulfate4-Methylaminophenol sulfate4-Methylaminophenol sulphate

A16236-0B A16236-30 A16236-18 A16236-18 A16236-30 A16236-0B
AAAA16236-18EA 29.99 USD InStock
AAAA16236-18 CAAAAA16236-18 AAAA16236-0B CAAAAA16236-0B AAAA16236-30 CAAAAA16236-30
4-(Methylamino)phenol sulfate 99%
4-(Methylamino)phenol sulfate


Formula: C₁₄H₂₀N₂O₆S
MW: 344.39 g/mol
Melting Pt: ca 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013140
CAS Number: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: "6.1",III
Merck Index: 13,06046
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty