Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cupferron 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ammonium 2-oxo-1-phenylhydrazinolate

A16551-14 A16551-14
AAAA16551-14EA 38.59 USD InStock
AAAA16551-14 CAAAAA16551-14
Cupferron 97+%
Cupferron
Reagent for determination of Ce, Cu, Fe, Sn, and Ti


Formula: C₆H₉N₃O₂
MW: 155.16
Melting Pt: 154 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00078422
CAS Number: 135-20-6
EINECS: 205-183-2
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty