Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-1-Aminoindan 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-1-Indanylamine

A16585-03 A16585-06 A16585-03 A16585-06
AAA16585-06EA 139.99 USD InStock
AAA16585-06 AAAA16585-03 CAAAAA16585-03 CAAAA16585-06
(±)-1-Aminoindan 99%
(±)-1-Aminoindan


Formula: C₉H₁₁N
MW: 133.19 g/mol
Boiling Pt: 225 °C
Melting Pt: 2 °C
Density: 1.038
Flash Pt: 94 °C(201 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003799
CAS Number: 34698-41-4
EINECS: 252-158-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty