Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methylcrotonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3,3-Dimethylacrylic acid3-Methylbut-2-enoic acid

A16865-14 A16865-14 A16865-22 A16865-22 A16865-36 A16865-36
AAAA16865-14EA 33.05 USD InStock
CAAAAA16865-14 AAAA16865-14 CAAAA16865-22 CAAAA16865-36 AAA16865-36 AAA16865-22
3-Methylcrotonic acid 98%
3-Methylcrotonic acid


Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 198…200 °C
Melting Pt: 66…68 °C
Density: 0.969
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004366
CAS Number: 541-47-9
EINECS: 208-782-7
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty