Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Aminolevulinic acid hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: δ-Aminolevulinic Acid Hydrochloride5-Amino-4-oxopentanoic acid hydrochloride5-Amino-4-ketovaleric acid hydrochloride

A16942-ME A16942-04 A16942-04 A16942-ME
AAAA16942-04EA 263 USD InStock
CAAAAA16942-04 CAAAA16942-ME AAAA16942-04 AAA16942-ME
5-Aminolevulinic acid hydrochloride 99%
5-Aminolevulinic acid hydrochloride


Formula: C5H10ClNO3
MW: 167.59 g/mol
Melting Pt: ca 156 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012869
CAS Number: 5451-09-2
EINECS: 226-679-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty