Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium dihydrogen orthophosphate 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Lithium phosphate monobasicLithium dihydrogenorthophosphateLithium dihydrogenphosphateLithium dihydrogen phosphate

A16987-30 A16987-30
AAAA16987-30EA 92.69 USD InStock
AAAA16987-30 CAAAAA16987-30
Lithium dihydrogen orthophosphate 97%
Lithium dihydrogen orthophosphate


Formula: H₂LiO₄P
MW: 103.93 g/mol
Melting Pt: >100 °C
Density: 2,461
Storage Temperature: Ambient
MDL Number: MFCD00016188
CAS Number: 13453-80-0
EINECS: 236-633-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty