Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tannic acid

Supplier: Thermo Scientific Chemicals
Synonyms: TanninGallotannin

A17022-0B A17022-30 A17022-30 A17022-0B
AAAA17022-30EA 60.24 USD InStock
CAAAAA17022-0B AAAA17022-30 AAAA17022-0B CAAAAA17022-30
Tannic acid
Tannic acid
Used for enzyme immobilization and protein adsorption


Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Melting Pt: ca 218 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2
Merck Index: 12,09221
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty