Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

A17061-03 A17061-03 A17061-14 A17061-06 A17061-06 A17061-14
AAAA17061-14EA 302.99 USD InStock
AAAA17061-14 CAAAAA17061-14 CAAAA17061-06 CAAAA17061-03 AAA17061-03 AAA17061-06
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole 98%
2-Amino-5-(trifluoromethyl)-1,3,4-thiadiazole


Formula: C₃H₂F₃N₃S
MW: 169.13 g/mol
Melting Pt: 222…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00003109
CAS Number: 10444-89-0
EINECS: 233-930-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty