Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide activated neutral Brockmann Grade II

Supplier: Thermo Scientific Chemicals

A17141-36 A17141-0E
AAAA17141-0EEA 206.99 USD InStock
AAAA17141-0E AAAA17141-36
Aluminium oxide activated neutral Brockmann Grade II
Aluminium oxide activated neutral


Formula: Al₂O₃
MW: 101.96 g/mol
Density: 3,97
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty