Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triisopropylchlorosilane 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triisopropylchlorosilane

A17376-18 A17376-30 A17376-09 A17376-18 A17376-30 A17376-09
AAAA17376-30EA 1113.99 USD InStock
AAAA17376-30 AAA17376-09 CAAAAA17376-30 AAA17376-18 CAAAA17376-09 CAAAA17376-18
Triisopropylchlorosilane 97+%
Triisopropylchlorosilane


Formula: C₉H₂₁ClSi
MW: 192.8 g/mol
Boiling Pt: 200 °C
Density: 0.908
Flash Pt: 62 °C(144 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009656
CAS Number: 13154-24-0
UN: 2987
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty