Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorostyrene 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Fluoro-2-vinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC) 0.1%

A17425-14 A17425-22 A17425-22 A17425-14 A17425-06 A17425-06
AAAA17425-22EA 1823.99 USD InStock
CAAAA17425-14 CAAAAA17425-22 CAAAA17425-06 AAAA17425-22 AAA17425-14 AAA17425-06
2-Fluorostyrene 98% stabilized
2-Fluorostyrene


Formula: C₈H₇F
MW: 122.14 g/mol
Boiling Pt: 29…30 °C/4 mm
Density: 1.020
Flash Pt: 35 °C(95 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013550
CAS Number: 394-46-7
EINECS: 206-896-1
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty