Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetramethylammonium hexafluorophosphate 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tetramethylammoniumhexafluorophosphate

A17467-36 A17467-14 A17467-22 A17467-22 A17467-14 A17467-36
AAAA17467-36EA 847.72 USD InStock
AAAA17467-36 AAAA17467-14 CAAAAA17467-22 CAAAAA17467-14 CAAAAA17467-36 AAAA17467-22
Tetramethylammonium hexafluorophosphate 99+%
Tetramethylammonium hexafluorophosphate


Formula: C₄H₁₂F₆NP
MW: 219.11 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012012
CAS Number: 558-32-7
EINECS: 209-194-3
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty