Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrasodium diphosphate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium pyrophosphateSodium diphosphateSodium pyrophosphate tetrabasicTetrasodium diphosphatetetra-Sodium pyrophosphate

A17546-30 A17546-30 A17546-0B A17546-0B
AAAA17546-30EA 54.21 USD InStock
AAAA17546-30 CAAAAA17546-0B AAAA17546-0B CAAAAA17546-30
Tetrasodium diphosphate 98%
tetra-Sodium diphosphate


Formula: Na₄O₇P₂
MW: 265.9 g/mol
Melting Pt: 988 °C
Density: 2,534
Storage Temperature: Ambient
MDL Number: MFCD00003513
CAS Number: 7722-88-5
EINECS: 231-767-1
Merck Index: 13,09312
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty