Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triisopropyl borate 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tri-i-propylborate

A17592-AE A17592-AE A17592-0F A17592-AP A17592-0F A17592-AP
AAAA17592-0FEA 500.99 USD InStock
AAAA17592-0F AAAA17592-AE CAAAAA17592-0F CAAAAA17592-AE AAAA17592-AP CAAAAA17592-AP
Triisopropyl borate 98+%
Triisopropyl borate


Formula: C₉H₂₁BO₃
MW: 188.07 g/mol
Boiling Pt: 139…141 °C
Melting Pt: –59 °C
Density: 0.815
Flash Pt: 17 °C(62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty