Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,3,4-Trichlorotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-(Chloromethyl)-3,4-dichlorobenzene 3,4-Dichlorobenzyl chloride 1,2-Dichloro-4-(chloromethyl)benzene

A17672-30 A17672-22 A17672-14 A17672-22 A17672-30 A17672-18 A17672-18 A17672-14
AAA17672-22EA 77.79 USD InStock
CAAAAA17672-18 CAAAA17672-22 AAA17672-22 CAAAA17672-14 AAAA17672-30 CAAAAA17672-30 AAA17672-14 AAAA17672-18
α,3,4-Trichlorotoluene 98%
α,3,4-Trichlorotoluene


Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 99…101 °C/2 mm
Melting Pt: –3 °C
Density: 1.416
Flash Pt: 155 °C(311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000906
CAS Number: 102-47-6
EINECS: 203-033-0
UN: 1760
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty