Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Glutamic acid monohydrate 99%

Supplier: Thermo Scientific Chemicals
Synonyms: DL-Glutamic Acid Monohydrate

A17719-14 A17719-22 A17719-22 A17719-36 A17719-14 A17719-36
AAAA17719-14EA 32.59 USD InStock
CAAAAA17719-22 AAAA17719-14 AAAA17719-36 AAAA17719-22 CAAAAA17719-14 CAAAAA17719-36
DL-Glutamic acid monohydrate 99%
DL-Glutamic acid monohydrate


Formula: C₅H₉NO₄·1H₂O
MW: 165.15 g/mol (147.13 anhy)
Melting Pt: ca 200 °C (dec.)
Density: 1,460
Storage Temperature: Ambient
MDL Number: MFCD00150703
EINECS: 210-522-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty