Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

m-Tolylacetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (3-Methylphenyl)acetic acid

A17743-30 A17743-09 A17743-18 A17743-30 A17743-18 A17743-09
AAA17743-09EA 38.99 USD InStock
AAA17743-09 AAA17743-18 CAAAA17743-09 CAAAAA17743-30 AAAA17743-30 CAAAA17743-18
m-Tolylacetic acid 99%
m-Tolylacetic acid


Formula: (C₆H₄(CH₃))CH₂COOH
MW: 150.18 g/mol
Boiling Pt: 120…123 °C (26 mmHg)
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00004340
CAS Number: 621-36-3
EINECS: 210-683-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty