Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2'-Thiodiacetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Thiodiacetic AcidThiodiglycolic acid

A17880-22 A17880-22 A17880-36 A17880-36
AAAA17880-36EA 236.99 USD InStock
CAAAAA17880-36 AAAA17880-36 AAAA17880-22 CAAAAA17880-22
2,2'-Thiodiacetic acid 98%
2,2'-Thiodiacetic acid


Formula: C₄H₆O₄S
MW: 150.16 g/mol
Melting Pt: 127…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00004359
CAS Number: 123-93-3
EINECS: 204-663-9
UN: 3261
ADR: "8",III
Merck Index: 12,09467
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty